ENA458427690 Enamine

1-(1-ethyl-5-methyl-1H-pyrazol-4-yl)ethan-1-one

  • MDL number MFCD03167209
  • Cas Number : 946658-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-228479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228479
Availability details for
Quantity
  • Availability
  • Add to Cart