ENA458427920 Enamine

1-[(4-fluorophenyl)methyl]-1H-1,2,4-triazol-3-amine

  • MDL number MFCD03422581
  • Cas Number : 832739-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-228708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228708
Availability details for
Quantity
  • Availability
  • Add to Cart