ENA458427994 Enamine

3,4-dimethylbenzohydrazide

  • MDL number MFCD01357033
  • Cas Number : 42596-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-228782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228782
Availability details for
Quantity
  • Availability
  • Add to Cart