ENA458428141 Enamine

N-[1-(propan-2-yl)piperidin-4-ylidene]hydroxylamine

  • MDL number MFCD04967217
  • Cas Number : 832741-00-1
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-228929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228929
Availability details for
Quantity
  • Availability
  • Add to Cart