ENA458428308 Enamine

3-(4-nitro-1H-pyrazol-1-yl)propanenitrile

  • MDL number MFCD02029339
  • Cas Number : 1002243-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-229097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229097
Availability details for
Quantity
  • Availability
  • Add to Cart