ENA458428341 Enamine

2-(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)ethan-1-amine

  • MDL number MFCD04967723
  • Cas Number : 925634-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-229130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229130
Availability details for
Quantity
  • Availability
  • Add to Cart