ENA458428356 Enamine

2-[5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

  • MDL number MFCD04967804
  • Cas Number : 1006348-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-229145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229145
Availability details for
Quantity
  • Availability
  • Add to Cart