ENA458428394 Enamine

[(1,5-dimethyl-1H-pyrazol-4-yl)methyl][(4-methoxyphenyl)methyl]amine

  • MDL number MFCD04968206
  • Cas Number : 1006476-52-3
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-229183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229183
Availability details for
Quantity
  • Availability
  • Add to Cart