ENA458428399 Enamine

N-[(1-methyl-1H-pyrazol-4-yl)methyl]aniline

  • MDL number MFCD04968265
  • Cas Number : 1006322-84-4
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-229188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229188
Availability details for
Quantity
  • Availability
  • Add to Cart