ENA458428525 Enamine

4-methoxy-3-[(2,4,5-trichlorophenoxy)methyl]benzaldehyde

  • MDL number MFCD04969111
  • Cas Number : 832739-71-6
  • Molecular Weight : 345.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-229314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229314
Availability details for
Quantity
  • Availability
  • Add to Cart