ENA458428555 Enamine

ethyl 2-(4-bromo-1H-pyrazol-1-yl)butanoate

  • MDL number MFCD04969148
  • Cas Number : 1005650-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-229344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229344
Availability details for
Quantity
  • Availability
  • Add to Cart