ENA458428653 Enamine

4-amino-N,1-diethyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD04969542
  • Cas Number : 1006485-32-0
  • Molecular Weight : 182.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-229442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229442
Availability details for
Quantity
  • Availability
  • Add to Cart