ENA458428835 Enamine

4-[(3-amino-4-bromo-1H-pyrazol-1-yl)methyl]benzohydrazide

  • MDL number MFCD02240790
  • Cas Number : 375837-72-2
  • Molecular Weight : 310.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-229624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229624
Availability details for
Quantity
  • Availability
  • Add to Cart