ENA458429004 Enamine

1-(difluoromethyl)-3-methyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD04970130
  • Cas Number : 957490-44-7
  • Molecular Weight : 230.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-229793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229793
Availability details for
Quantity
  • Availability
  • Add to Cart