ENA458429010 Enamine

1-[5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine

  • MDL number MFCD04970170
  • Cas Number : 1006456-87-6
  • Molecular Weight : 233.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-229799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229799
Availability details for
Quantity
  • Availability
  • Add to Cart