ENA458429081 Enamine

4-[2-(morpholin-4-yl)ethoxy]aniline

  • MDL number MFCD04970981
  • Cas Number : 52481-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-229870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-229870
Availability details for
Quantity
  • Availability
  • Add to Cart