ENA458429380 Enamine

2-(3-methyl-1H-pyrazol-1-yl)ethan-1-amine

  • MDL number MFCD01822277
  • Cas Number : 62821-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-230170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230170
Availability details for
Quantity
  • Availability
  • Add to Cart