ENA458429462 Enamine

5-methyl-1-(2-methylpropyl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD06805461
  • Cas Number : 1006496-01-0
  • Molecular Weight : 236.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-230252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230252
Availability details for
Quantity
  • Availability
  • Add to Cart