ENA458429792 Enamine

2-(5-bromo-4-formyl-2-methoxyphenoxy)ethyl acetate

  • MDL number MFCD06631001
  • Cas Number : 836662-18-1
  • Molecular Weight : 317.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-230582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230582
Availability details for
Quantity
  • Availability
  • Add to Cart