ENA458429799 Enamine

3-bromo-4-(carbamoylmethoxy)-5-ethoxybenzoic acid

  • MDL number MFCD05996129
  • Cas Number : 713104-22-4
  • Molecular Weight : 318.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-230589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230589
Availability details for
Quantity
  • Availability
  • Add to Cart