ENA458429892 Enamine

4-methyl-5-[3-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-2-amine

  • MDL number MFCD08558209
  • Cas Number : 1006348-68-0
  • Molecular Weight : 248.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-230682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230682
Availability details for
Quantity
  • Availability
  • Add to Cart