ENA458429916 Enamine

N-(5-acetyl-4-methyl-1,3-thiazol-2-yl)-N-cyclopropyl-4-methylbenzamide

  • MDL number MFCD08554858
  • Cas Number : 937598-04-4
  • Molecular Weight : 314.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-230706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230706
Availability details for
Quantity
  • Availability
  • Add to Cart