ENA458429920 Enamine

3-(trifluoromethyl)-1H,4H,5H,6H-cyclopenta[c]pyrazole

  • MDL number MFCD04973035
  • Cas Number : 347361-46-0
  • Molecular Weight : 176.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-230710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230710
Availability details for
Quantity
  • Availability
  • Add to Cart