ENA458430167 Enamine

4-(4-methylpiperazin-1-yl)benzonitrile

  • MDL number MFCD03546779
  • Cas Number : 34334-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-230957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230957
Availability details for
Quantity
  • Availability
  • Add to Cart