ENA458430207 Enamine

1-{1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl}piperidine-4-carboxylic acid

  • MDL number MFCD08559087
  • Cas Number : 937600-33-4
  • Molecular Weight : 261.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-230997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-230997
Availability details for
Quantity
  • Availability
  • Add to Cart