ENA458430282 Enamine

2-[4-(difluoromethyl)-6-(3-methoxyphenyl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

  • MDL number MFCD08697733
  • Cas Number : 937607-17-5
  • Molecular Weight : 347.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231072
Availability details for
Quantity
  • Availability
  • Add to Cart