ENA458430376 Enamine

2-methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine

  • MDL number MFCD08559043
  • Cas Number : 1006348-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-231166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231166
Availability details for
Quantity
  • Availability
  • Add to Cart