ENA458430418 Enamine

1-(2,2-difluoroethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD08559011
  • Cas Number : 1006959-46-1
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231208
Availability details for
Quantity
  • Availability
  • Add to Cart