ENA458430508 Enamine

2-methyl-1-[(4-methylphenyl)methyl]-1H-indole-3-carbaldehyde

  • MDL number MFCD03906282
  • Cas Number : 592550-44-2
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231298
Availability details for
Quantity
  • Availability
  • Add to Cart