ENA458430511 Enamine

1-(2,2-difluoroethyl)-3-methyl-1H-pyrazole

  • MDL number MFCD09473545
  • Cas Number : 1171013-54-9
  • Molecular Weight : 146.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231301
Availability details for
Quantity
  • Availability
  • Add to Cart