ENA458430594 Enamine

1-(4-fluorophenyl)-3-(propan-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD09473357
  • Cas Number : 1154199-53-7
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231384
Availability details for
Quantity
  • Availability
  • Add to Cart