ENA458430850 Enamine

N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]cyclopropanamine

  • MDL number MFCD09701801
  • Cas Number : 1170202-73-9
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231640
Availability details for
Quantity
  • Availability
  • Add to Cart