ENA458430869 Enamine

1-[(3-cyclopropyl-1-methyl-1H-pyrazol-5-yl)methyl]piperidin-4-amine

  • MDL number MFCD09701820
  • Cas Number : 1171769-79-1
  • Molecular Weight : 234.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231659
Availability details for
Quantity
  • Availability
  • Add to Cart