ENA458430923 Enamine

1-methyl-5-(pyrrolidin-2-yl)-1H-pyrazole

  • MDL number MFCD09701945
  • Cas Number : 1171787-08-8
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231713
Availability details for
Quantity
  • Availability
  • Add to Cart