ENA458430925 Enamine

1-methyl-3-(pyrrolidin-2-yl)-1H-pyrazole

  • MDL number MFCD09701947
  • Cas Number : 1169954-20-4
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231715
Availability details for
Quantity
  • Availability
  • Add to Cart