ENA458431040 Enamine

3-cyclopropyl-6-methyl-1-phenyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09834255
  • Cas Number : 1018127-41-7
  • Molecular Weight : 293.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231830
Availability details for
Quantity
  • Availability
  • Add to Cart