ENA458431088 Enamine

6-chloro-4-(difluoromethyl)-2-methyl-2H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09834310
  • Cas Number : 1018165-42-8
  • Molecular Weight : 217.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-231878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-231878
Availability details for
Quantity
  • Availability
  • Add to Cart