ENA458431237 Enamine

3-[7-(thiophen-2-yl)-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl]propanoic acid

  • MDL number MFCD10001595
  • Cas Number : 1018052-13-5
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-232027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-232027
Availability details for
Quantity
  • Availability
  • Add to Cart