ENA458431270 Enamine

6-ethyl-1-(2-fluorophenyl)-3-methyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD10699000
  • Cas Number : 1119499-72-7
  • Molecular Weight : 299.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-232060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-232060
Availability details for
Quantity
  • Availability
  • Add to Cart