ENA458431699 Enamine

1,3-dimethyl-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD15976435
  • Cas Number : 1245820-90-9
  • Molecular Weight : 194.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-232475

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-232475
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart