ENA458431995 Enamine

4-iodo-1,5-dimethyl-3-nitro-1H-pyrazole

  • MDL number MFCD24539555
  • Cas Number : 1354705-66-0
  • Molecular Weight : 267.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-232772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-232772
Availability details for
Quantity
  • Availability
  • Add to Cart