ENA458432027 Enamine

1-methyl-4-(thiophen-2-yl)-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid

  • MDL number MFCD24540302
  • Cas Number : 1356543-40-2
  • Molecular Weight : 259.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-232804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-232804
Availability details for
Quantity
  • Availability
  • Add to Cart