ENA458432100 Enamine

1-ethyl-5-fluoro-4-iodo-1H-pyrazole

  • MDL number MFCD24540499
  • Cas Number : 1392274-31-5
  • Molecular Weight : 240.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-232877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-232877
Availability details for
Quantity
  • Availability
  • Add to Cart