ENA458432117 Enamine

3-iodo-1-methyl-4-nitro-1H-pyrazole

  • MDL number MFCD25371333
  • Cas Number : 186298-52-2
  • Molecular Weight : 253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-232894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-232894
Availability details for
Quantity
  • Availability
  • Add to Cart