ENA458432275 Enamine

1-cyclopentyl-3-nitro-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD28155641
  • Cas Number : 1795473-86-7
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-233052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-233052
Availability details for
Quantity
  • Availability
  • Add to Cart