ENA458432431 Enamine

2-methyl-2-[4-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD28962904
  • Cas Number : 1823464-45-4
  • Molecular Weight : 236.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-233215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-233215
Availability details for
Quantity
  • Availability
  • Add to Cart