ENA458434178 Enamine

6-methyl-3-(methylsulfanyl)-4,5-dihydro-1,2,4-triazin-5-one

  • MDL number MFCD00711682
  • Cas Number : 1566-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-235970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-235970
Availability details for
Quantity
  • Availability
  • Add to Cart