ENA458435516 Enamine

3-(1,3-dioxaindan-5-yl)-5-(chloromethyl)-1,3-oxazolidin-2-one

  • MDL number MFCD16631777
  • Cas Number : 1204298-23-6
  • Molecular Weight : 255.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-237306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-237306
Availability details for
Quantity
  • Availability
  • Add to Cart