ENA458436721 Enamine

2-(3-oxo-1,3-dihydro-2,1-benzothiazol-1-yl)acetic acid

  • MDL number MFCD09743284
  • Cas Number : 55114-02-8
  • Molecular Weight : 209.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-238515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-238515
Availability details for
Quantity
  • Availability
  • Add to Cart