ENA458437536 Enamine

2-(5-amino-3-methyl-1H-pyrazol-1-yl)-6-propyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD11986605
  • Cas Number : 1171768-45-8
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-239330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-239330
Availability details for
Quantity
  • Availability
  • Add to Cart